Order Entry
ContactUsLinkComponent
Anti-NPYY2 Rabbit Polyclonal Antibody
Anti-NPYY2 Rabbit Polyclonal Antibody
Catalog #: 89368-024
Supplier:  Genetex
Anti-NPYY2 Rabbit Polyclonal Antibody
Catalog #: 89368-024
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Neuropeptide Y Y2
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Antigen symbol:
    NPYY2
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Storage temperature:
    Upon Receipt - Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Tested applications:
    IHC
  • Cat. no.:
    89368-024
  • Supplier No.:
    GTX80744

Specifications

About this item

This NPY Y2 antibody has been extensively characterized at the spinal level and is currently being characerized in brain.? The antibody has been tested in both rat and mouse and appears to work better on mose tissue, although positive staining is seen in rat.? For full characterizarion in dorsal root ganglion and spinal cord see: J Comp Neurol. 2005 Aug 29;489(3):328-48.

Type: Primary
Antigen: NPYY2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Customers Who Bought This Also Bought