Order Entry
ContactUsLinkComponent
Anti-ITGA2 Mouse Monoclonal Antibody [clone: AK7]
Catalog #: 89366-114
Supplier:  Genetex
Price
$810.84
Log in for availability
Unit of Measure Each (100µG)
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Integrin Alpha 2
  • Clonality:
    Monoclonal
  • Clone:
    AK7
  • Gene ID:
    3673
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Baboon
  • Antigen symbol:
    ITGA2
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • Antigen synonyms:
    BR
    GPIa
    VLAA2
    DX5 antigen
    pan-NK cells
    CD49B
    HPA-5
    VLA-2
    ITGA2
  • Storage buffer:
    Phosphate buffered saline containing 0.09% Sodium Azide
  • Storage temperature:
    Store at +4 °C or at –20 °C if preferred. This product should be stored undiluted. Storage in frost-free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
  • Tested applications:
    Fluorescence-Activated Cell Sorting, Functional Assay
  • Purification:
    Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
  • Cat. no.:
    89366-114
  • Size:
    100 μg

Specifications

About this item