To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Bromodomain Containing 2
- Antigen symbol:BRD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:FSH|RING3|FSRG1|O27.1.1|Bromodomain-containing protein 2|D6S113E|NAT|RING3 protein|RNF3
- Immunogen:Synthetic peptide representing a portion of the protein between the putative nuclear localization signal and the ET domain (LocusLink ID 6046).
- Purification:Affinity purified with antigen
- Storage buffer:0.1ml Tris citrate/phosphate, pH 7-8, containing 0.1% sodium azide
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Tested applications:WB
- Cat. no.:89367-050
- Supplier No.:GTX19276
Specifications
About this item
BRD2 is a putative nuclear transcriptional regulator and a member of a family of genes that are expressed during development.
Type: Primary
Antigen: BRD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human