Order Entry
ContactUsLinkComponent
Anti-MMP10 Rabbit Polyclonal Antibody
Anti-MMP10 Rabbit Polyclonal Antibody
Catalog # 89366-270
Supplier:  Genetex
Anti-MMP10 Rabbit Polyclonal Antibody
Catalog # 89366-270
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 10
  • Antigen symbol:
    MMP10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 μg
  • Antigen synonyms:
    transin-2|matrix metalloproteinase-10|stromelysin 2|stromelysin-2|MMP-10|matrix metalloprotease 10|transin 2|matrix metalloproteinase 10 (stromelysin 2)
  • Gene ID:
    4319
  • Concentration:
    1 mg/ml
  • Immunogen:
    Polyclonal antibody produced in rabbits immunized with a synthetic peptide corresponding to a region of human MMP10
  • Purification:
    Protein A/G affinity purified
  • Storage buffer:
    PBS with 2% sucrose
  • Storage temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Tested applications:
    WB
  • Cat. no.:
    89366-270
  • Supplier No.:
    GTX77732

Specifications

About this item

Rabbit Polyclonal Antibody to MMP10

WB: Suggested dilution at 1 ?g/ml in 5% skim milk / PBS buffer. Optimal dilutions should be determined by end-user.

Type: Primary
Antigen: MMP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Frequently Bought Together