Order Entry
ContactUsLinkComponent
Anti-HLA-G Mouse Monoclonal Antibody [clone: MEM-G/9]
Anti-HLA-G Mouse Monoclonal Antibody [clone: MEM-G/9]
Catalog #: 89366-598
Supplier:  Genetex
Anti-HLA-G Mouse Monoclonal Antibody [clone: MEM-G/9]
Catalog #: 89366-598
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HLA class I histocompatibility antigen, alpha chain G
  • Clonality:
    Monoclonal
  • Clone:
    MEM-G/9
  • Gene ID:
    3135
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Antigen symbol:
    HLA-G
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    100 μg
  • Western blot:
    Yes
  • Antigen synonyms:
    HLA class I histocompatibility antigen|DADB-15K14.8|alpha chain G|HLA-G histocompatibility antigen|b2 microglobulin|MHC-G|HLA class I molecule|HLA G antigen|MHC class I antigen G|class I|G
  • Storage buffer:
    Azide free phosphate buffered saline (PBS), preserved by filter sterilization, approx. pH 7.4
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Recombinant human HLA-G refolded with beta2-microglobulin and peptide.
  • Tested applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    > 95% (by SDS-PAGE)
  • Cat. no.:
    89366-598
  • Supplier No.:
    GTX78332

Specifications

About this item

Monoclonal Antibody to HLA-G (Human) Azide free

FACS: Recommended dilution: 2-5 ug/ml. Positive control: JEG-3 human choriocarcinoma cell line.ELISA: Coating antibody (10 ug/ml) Detection antibody (biotin or peroxidase conjugate (1 ug/ml)

Type: Primary
Antigen: HLA-G
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Customers Who Bought This Also Bought