Anti-IgG Sheep Polyclonal Antibody
1 mg
Catalog # 89362-212
Supplier: Genetex
Supplier #: GTX26867
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Rhodamine-conjugated Sheep anti-Human IgG polyclonal antibody
Recommended Starting Dilutions: Not specifiedOptimal titers for applications should be determined by the researcher.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Specifications
- Size:1 mg
- Clonality:Polyclonal
- Flow cytometry:Yes
- Purification:IgG fraction
- Antibody type:Primary
- Isotype:IgG
- Storage temperature:Keep as concentrated solution. For short-term storage, store at 4 °C. For long-term storage, aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles. Protect from light.
- Host:Sheep
- Concentration:2 mg/ml
- Antigen name:Immunoglobulin G
- Cat. No.:89362-212
- Reactivity:Human
- Immunogen:Human IgG whole molecule
- ImmunoChemistry:Yes
- Conjugation:Rhodamine
- Tested applications:Fluorescence-Activated Cell Sorting, Immunomicroscopy
- Antigen symbol:IgG
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 , 10 mg/mL Bovine Serum Albumin (BSA) , 0.01% (w/v) Sodium Azide
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



