Order Entry
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody
Anti-IgG Goat Polyclonal Antibody
Catalog # 89362-368
Supplier:  Genetex
Anti-IgG Goat Polyclonal Antibody
Catalog # 89362-368
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Immunogen:
    Full length native Mouse IgG (purified).
  • Purification:
    IgG fraction
  • Storage buffer:
    0.05M Tris Chloride, 0.15M sodium chloride, 0.001M magnesium chloride, 0.0001M zinc chloride, 50% glycerol; pH 8.0, 10 mg/ml bovine serum albumin, containing 0.01% sodium azide
  • Storage temperature:
    Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
  • Tested applications:
    Fluorescence-Activated Cell Sorting, Immunomicroscopy
  • Cat. no.:
    89362-368
  • Supplier No.:
    GTX27064

Specifications

About this item

Immunoglobulin G (IgG) is one of the most abundant protein in serum with normal levels between 8-17 mg/ml in adult blood. IgG is important for our defense against microorganisms and the molecules are produced by B-lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions: to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse

Frequently Bought Together