Anti-5HT2C Rabbit Polyclonal Antibody
89366-434
: Genetex
- Antibody type:Primary
- Antigen name:5-hydroxytryptamine receptor 2C
- Antigen symbol:5HT2C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:25 μg
- Gene ID:15560
- Antigen synonyms:5-HT2cR|SR1|15560|5HT2cR|Htr2c|5HT|5HT2C receptor|Htr1c|5 HT2cR|5 HT2C receptor|5HT1c
- Storage buffer:Phosphate buffered saline, pH 7.7, containing 0.01% sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide - KLH conjugated. Peptide sequence identity/similarity to Mouse: 55/65%. Rat: 60/70%.
- Tested applications:IHC-P
- Purification:Peptide immunogen affinity column
- Cat. no.:89366-434
- Supplier No.:GTX77939
Rabbit polyclonal to 5-HT2C Receptor
Type: Primary
Antigen: 5HT2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human