Chromatography Column, 1:5 PTFE Plug, #15 Ace-Thred, Ace Glass Incorporated
This chromatography column has a #15 Ace-Thred at top and a 2 mm bore 1:5 taper solid PTFE stopcock plug at the bottom.
Fabricated from borosilicate glass.
: Complete item consists of column with 89059-220 bushing and O-Ring, and a 89052-938 tubing connector for connecting 12.5 to 14 mm ext.Ø tubing to column. Components may also be purchased separately. NOTE: when using with 89052-938 tubing connector, the O-Ring that is supplied with the bushing is NOT necessary.
Frequently Bought Together