ContactUsLinkComponent
49 results for Tongs
You searched for: Tongs
Tongs
Provide maximum personnel safety when transferring corrosive or extreme temperature-subjected samples by using laboratory tongs. The pinching, concave arms operate similarly to scissors and have a strong metal construction for the chemical and temperature resistance required. Uniquely shaped prongs accommodate different specimen containers and may have coated tips for additional security when moving hazardous items. Crucial in testing environments, tongs also guarantee that inaccuracies in measurements do not occur.
Sort by
VWR® Crucible Tongs, Julian
VWR® Crucible Tongs, Julian
Supplier: VWR International
Stainless steel Julian-style crucible tongs with compressing guide.
Expand 1 Items
Loading...
VWR® Flask Tongs
VWR® Flask Tongs
Supplier: VWR International
VWR® Flask Tongs are made from ASTM Standard (899-94) Series 400 Stainless Steel.
Expand 1 Items
Loading...
VWR® Tongs, PTFE-Coated
VWR® Tongs, PTFE-Coated
Supplier: VWR International
Designed to handle hot, cold, corrosive, or sticky materials without scratching or damaging container exteriors.
Expand 1 Items
Loading...
VWR® Tongs, Locking Joint
VWR® Tongs, Locking Joint
Supplier: VWR International
Tongs are constructed of electropolished stainless steel and feature a locking joint.
Expand 1 Items
Loading...
VWR® Jumbo Crucible Tongs
VWR® Jumbo Crucible Tongs
Supplier: VWR International
VWR® Jumbo Crucible Tongs are made from ASTM Standard (899-94) Series 400 Stainless Steel.
Expand 1 Items
Loading...
Crucible Tongs
Crucible Tongs
Supplier: VWR International
Crucible Tongs
Expand 2 Items
Loading...
VWR® Beaker Tongs
VWR® Beaker Tongs
Supplier: VWR International
Tongs feature a three-prong design and carbon steel, cadmium-plated construction.
Expand 1 Items
Loading...
VWR® Tongs, Riveted Joint
VWR® Tongs, Riveted Joint
Supplier: VWR International
Tongs are constructed of electropolished stainless steel and feature a riveted joint.
Expand 1 Items
Loading...
VWR® Replacement Fiberglass Sleeve for Beaker Tongs
VWR® Replacement Fiberglass Sleeve for Beaker Tongs
Supplier: VWR International
Replacement fiberglass sleeve covered jaws are designed for use with beaker tongs.
Expand 1 Items
Loading...
VWR® Dish Tongs
VWR® Dish Tongs
Supplier: VWR International
VWR® Dish Tongs are made from ASTM Standard (899-94) Series 400 Stainless Steel.
Expand 1 Items
Loading...
Beaker Tongs
Beaker Tongs
Supplier: VWR International
These long-lasting beaker tongs are made of nickel-plated steel.
Expand 2 Items
Loading...
VWR® Crucible Tongs with PTFE-Coated Tips
VWR® Crucible Tongs with PTFE-Coated Tips
Supplier: VWR International
VWR® Crucible Tongs with PTFE Coated Tips are made from ASTM Standard (899-94) Series 400 Stainless Steel.
Expand 1 Items
Loading...
VWR® Stainless Steel Crucible Holders, Corrugated Tip
VWR® Stainless Steel Crucible Holders, Corrugated Tip
Supplier: VWR International
VWR® Stainless Steel Crucible Holders have tight-fitting corrugated tips for handling dishes in muffle furnaces.
Expand 1 Items
Loading...
VWR® Crucible Tongs
VWR® Crucible Tongs
Supplier: VWR International
Crucible tongs are designed for use in handling a wide variety of laboratory apparatus.
Expand 2 Items
Loading...
VWR® Beaker Tongs, Fiberglass Coating
VWR® Beaker Tongs, Fiberglass Coating
Supplier: VWR International
Tongs feature a stainless steel construction, a riveted joint, and fiberglass coating for use in high-temperature applications.
Expand 1 Items
Loading...
VWR® Platinum Tip Crucible Tongs
VWR® Platinum Tip Crucible Tongs
Supplier: VWR International
VWR® Platinum Tip Crucible Tongs provide a contamination-free gripping surface.
Expand 1 Items
Loading...
VWR® Beaker Tongs, Plastic-Coated Tips
VWR® Beaker Tongs, Plastic-Coated Tips
Supplier: VWR International
Tongs feature carbon steel, cadmium-plated construction with plastic-coated tips.
Expand 1 Items
Loading...
Holder for Sample Tubes, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Expand 1 Items
Loading...
Cryo Tongs, Thermo Scientific
Supplier: Thermo Fisher Scientific
Ideal for the safe retrieval of cryopreservation storage products.
Expand 1 Items
Loading...
SP Bel-Art Stainless Steel Tongs, Bel-Art Products, a part of SP
SP Bel-Art Stainless Steel Tongs, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Stainless steel, laboratory type tongs have 7.6 cm (3") tips coated with brown Teflon® FEP.
Expand 1 Items
Loading...
SP Bel-Art Sterileware® Sterile Mini Tongs, Bel-Art Products, a part of SP
SP Bel-Art Sterileware® Sterile Mini Tongs, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
These small, lightweight tongs are ideal for sampling small quantities of material ranging from food to pharmaceuticals.
Expand 1 Items
Loading...
Crucible Tongs, Plated Steel, United Scientific Supplies
Crucible Tongs, Plated Steel, United Scientific Supplies
Supplier: United Scientific Supplies
Tongs have an overall length of 9" and include riveted joints and serrated tips.
Expand 1 Items
Loading...
Scorifier Steel Tongs, Webber's Welding
Supplier: Webber's Welding
The scorifier tongs are made of mild steel for dependable, long-lasting use.
Expand 1 Items
Loading...
Crucible Tongs with Straight Tips
Crucible Tongs with Straight Tips
Supplier: Wards
These metal tongs feature a bow design and straight serrated tips. They provide a secure grip for crucibles in laboratory settings.
Expand 1 Items
Loading...
Crucible Tongs
Crucible Tongs
Supplier: United Scientific Supplies
Hold crucibles at a safe distance.
Expand 1 Items
Loading...
Heavy-Duty Beaker Tongs
Heavy-Duty Beaker Tongs
Supplier: Wards
These 12" long stainless steel tongs with cork-lined jaws and a riveted joint are ideal for handling hot beakers ranging from 100 to 1500 ml capacities.
Expand 1 Items
Loading...
Chemware® Tongs, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
Chemware® PTFE Fluoropolymer Coated Tongs are durable stainless steel tongs covered with an extra-heavy, non-stick PTFE coating that will not scratch or damage delicate surfaces.
Expand 1 Items
Loading...
Flask Tongs, Stainless Steel, United Scientific Supplies
Flask Tongs, Stainless Steel, United Scientific Supplies
Supplier: United Scientific Supplies
Stainless steel flask tongs are 12” long with riveted joints.
Expand 1 Items
Loading...
Sampling Tongs
Sampling Tongs
Supplier: Wards
These nickel-plated tongs are ideal for sampling purposes, providing a reliable and precise grip for various materials in laboratory and industrial environments.
Expand 1 Items
Loading...
Crucible Tongs, Steel, Nickel, Webber's Welding
Supplier: Webber's Welding
These crucible tongs are made of mild steel for durable and dependable service.
Expand 1 Items
Loading...
Sort by
Recommended for You