You searched for: TLC Plates
TLC plates are crucial in analyzing mixtures through compound separation to identify the quantity and purity. Product appearances or reactant disappearances are easily observed to show reaction progress. Pre-coated with a thin, uniform layer of solid absorbent, the TLC plates support accurate and reproducible conclusions. Choose from standard sized glass, metal, or plastic constructions compatible with manual or automated testing. Pore sizes may vary among models. Select styles may feature fluorescent indicator for colorless compounds.
Baker-flex® Precoated Flexible TLC Sheets, J.T. Baker®, Avantor
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Ready-to-use in thin layer chromatography applications, Baker-flex® precoated flexible plastic sheets have a hard, abrasion-resistant coating and solvent-resistant substrate for rapid, clean separations.
Expand 22 Items
Precoated Glass-Backed TLC Plates, MilliporeSigma
Supplier: MilliporeSigma
For analytical TLC, high-performance analytical TLC, or preparative TLC
Expand 77 Items
TLC Plates, Silica Gel 60 F254 MS-grade, MilliporeSigma
Supplier: MilliporeSigma
These TLC plates are coated with TLC silica gel with flourescent indicator F254 and are ideal for thin layer chromatography applications.
Expand 1 Items
TLC Glass Plates, Silica Gel 60, MilliporeSigma
Supplier: MilliporeSigma
These new TLC silica gel 60G plates are recommended for customers in QA/QC labs using older Ph Eur monograph methods, which require TLC plates with gypsum binder.
Expand 2 Items
TLC Silica Gel 60 Plates and Sheets, MilliporeSigma
Supplier: MilliporeSigma
TLC plates are combination of Millipore silica gel 60 and a unique polymeric binder resulting in adherent and hard surface.
Expand 4 Items
Glass Chromatography Plates, MilliporeSigma
Supplier: MilliporeSigma
Tlc Luxplate* Silica Gel 60 F254 50 Glass Plates 10 X 20 Cm
Expand 2 Items
Aluminium Oxide F254 Plates And Sheets, MilliporeSigma
Supplier: MilliporeSigma
Aluminium oxide plates are used to analyze basic and neutral compounds at different pH levels
Expand 2 Items
Precoated Plastic-Backed TLC Sheets, MilliporeSigma
Supplier: MilliporeSigma
For analytical TLC
Expand 6 Items
Plates, Laser Coded, Silica Gel 60, MilliporeSigma
Supplier: MilliporeSigma
These plates feature individual laser coding for GLP applications
Expand 3 Items
PLC Plates, MilliporeSigma
Supplier: MilliporeSigma
PLC plates allow users to separate samples that vary greatly in size - from grams down to milligrams
Expand 1 Items
Precoated Aluminum-Backed TLC Sheets, MilliporeSigma
Supplier: MilliporeSigma
For analytical TLC or high-performance analytical TLC
Expand 21 Items
RP Modified TLC Silica Gel 60 Sheets and Plates, MilliporeSigma
Supplier: MilliporeSigma
RP-modified plates serve two purposes: Act as a pilot method for HPLC and allow to choose various solvent system for special separations
Expand 2 Items
Multiformat Pre-Scored Plates, Silica Gel 60 F254, MilliporeSigma
Supplier: MilliporeSigma
These pre-scored glass plates fold into smaller formats with just one easy snap of the fingers.
Expand 1 Items
HPTLC Silica Gel 60 Plates and Sheets, MilliporeSigma
Supplier: MilliporeSigma
HPTLC plates deliver fast and quantitative analysis of complex samples for manual or automated use.
Expand 6 Items
HPTLC Silica Gel 60 Plates and Sheets, MS-Grade, MilliporeSigma
Supplier: MilliporeSigma
HPTLC silica gel 60, Fluorescent indicator: F254 MS-grade, Glass Plate, Plate size: 20 x 10 cm, Layer thickness: 200 um, Pore size: 60 armstrong
Expand 3 Items
HPTLC Silica Gel 60 AMD Plates, Extra Thin, MilliporeSigma
Supplier: MilliporeSigma
HPTLC Plates AMD with extra thin layer of only 100 µm have been specially developed for even more demanding application such as automated multiple development (AMD)
Expand 1 Items
Thin Layer Plates And Sheets, LiChrospher® Silica Gel 60, MilliporeSigma
Supplier: MilliporeSigma
Unique HPTLC Lichrospher® plates are the first thin layer chromatography plates based on spherical silica particles
Expand 2 Items
Preparative TLC Plates, Agela Technologies
Supplier: Agela Technologies
These thin layer chromatography plates are used for a variety of chemistries
Expand 1 Items
TLC Plates, Special Layers, Glass Backing, Chiral Plate, Macherey-Nagel
Supplier: Macherey-Nagel
These glass plates with a special layer are used for TLC enantiomer separations (e.g. amino acids, thiazolidine derivatives, dipeptides, lactones, α-hydroxycarboxylic acids).
Expand 1 Items
Silica Gel GHLF UNIPLATES™ TLC Plates, Analtech, Miles Scientific
Supplier: Analtech
These plates are 100% water resistant and are compatible with aqueous developing solvents for efficient separations.
Expand 2 Items
HPTLC Plates, Nano Silica Gel 60, Unmodified Layers, Nano-SIL, Macherey Nagel
Supplier: Macherey-Nagel
Unmodified HPTLC silica layers for HPTLC with nano silica gel (particle size 2 to 10 μm).
Expand 2 Items
Preparative TLC Plates, Analtech
Supplier: Analtech
Preparative TLC plates feature thicker adsorbent layers and are useful for separation of larger quantities of material than plates with a 250 µm thick coating.
Expand 2 Items
TLC Plates, Cellulose MN 300, Macherey-Nagel
Supplier: Macherey-Nagel
Fibrous cellulose layers are available as POLYGRAM® polyester sheets.
Expand 2 Items
TLC Standard Silica Plate, Phenomenex
Supplier: Agela Technologies
Standard sized thin-layer chromatography plate, with standard silica treatment and aluminum back
Expand 1 Items
Thin Layer Chromatography Plates, SiliCycle
Supplier: SiliCycle
SiliaPlate™ TLC Plates are an optimal choice for quick, inexpensive, versatile and classical separations, that can purposefully be functionalized for extra selectivity toward your analyte.
Expand 37 Items
TLC Plates, Silica Gel 60, Unmodified Layers, Polyester Sheets, SIL G, POLYGRAM®, Macherey-Nagel
Supplier: Macherey-Nagel
Ready-to-use sheets
Expand 2 Items
HPTLC Plates, Silica Gel 60, Modified Layers, RP-18 W/UV₂₅₄, Macherey Nagel
Supplier: Macherey-Nagel
Octadecyl-modified HPTLC silica layers are available as glass plates.
Expand 2 Items
TLC Plate, Silica, Phenomenex
Supplier: Agela Technologies
These TLC Silica Plates are glass-backed and serve as a tool for reaction monitoring and method development
Expand 3 Items
UNIPLATES™ Hard Layer Silica Gel HLF Plates, Analtech
Supplier: Analtech
Hard layer silica gel HLF UNIPLATES™ (Organic Binder) are the most rugged silica gel plates. They not only ease handling and sample application, but also permit the use of up to 80% water in the developing solvent without loss of adherence of the adsorbent layer to the glass plate.
Expand 2 Items
Silica Gel TLC Plates with Gypsum Binder, Analtech, Miles Scientific
Supplier: Analtech
These unique plates include calcium sulfate hemihydrate (CaSO41/2H2O), a refined form of gypsum, in the adsorbent formulation