Order Entry
ContactUsLinkComponent
133 results for Tubes

You searched for: Test Tubes

Test Tubes

High-quality test tubes are designed and constructed for optimal performance and excellent resistance to chemicals. Sturdy construction from borosilicate glass delivers a great range of operating temperatures. These precision-molded test and sample tubes are suited to a wide variety of applications, and are available in round- or flat-bottomed models. Easy-to-read graduations and labeling with all models. Minimal thermal expansion gives these test tubes high resistance to temperature changes.

Sort by
VWR® Test Tubes with Sturdy Lip, Borosilicate Glass

VWR® Test Tubes with Sturdy Lip, Borosilicate Glass

Supplier: VWR International

High quality test tubes designed and constructed for optimal performance.

Expand 9 Items
Loading...
VWR® Test Tubes

VWR® Test Tubes

Supplier: VWR International

VWR® Test Tubes

Expand 9 Items
Loading...

Ward's® Test Tube Homeschool Pack

Supplier: Avantor

Test tubes and everything you need to use them.

Expand 1 Items
Loading...
Avantor OmniTop Assemblies, Pre-Sterilized Single-Use Sample Tubes®, Avantor Fluid Handling

Avantor OmniTop Assemblies, Pre-Sterilized Single-Use Sample Tubes®, Avantor Fluid Handling

Supplier: Avantor Fluid Handling

OmniTop Sample Tubes® are a convenient device that can be used to obtain fluid samples. Each single-use OmniTop tube comes with a pre-attached 0.2 µm vent filter and 18" of tubing (C-Flex®, TYGON®, silicone or PharMed®).

Expand 10 Items
Loading...
Expand 1 Items
Loading...
Test Tube, Graduated with Beaded Rim, Ace Glass Incorporated

Test Tube, Graduated with Beaded Rim, Ace Glass Incorporated

Supplier: Ace Glass

This is a graduated beaded rim test tube for standard laboratory use.

Expand 2 Items
Loading...
Test Tubes, Ace Glass Incorporated

Test Tubes, Ace Glass Incorporated

Supplier: Ace Glass

Test tubes, available with or without matching glass stoppers; [ST] 24/40 stopper is hollow; #16 and #19 stoppers are solid.

Expand 8 Items
Loading...
Quartz Test Tube Without Lip, Ace Glass Incorporated

Quartz Test Tube Without Lip, Ace Glass Incorporated

Supplier: Ace Glass

This tube is made from quartz.

Expand 1 Items
Loading...
KONTES® Soil Analysis Tubes, Kimble Chase

KONTES® Soil Analysis Tubes, Kimble Chase

Supplier: DWK Life Sciences (KIMBLE)

These test tubes are designed for use with Tekmar® 2016/2032 autosamplers and 4100/4200 automatic samplers equipped with 1.9 cm (³/₄") mounts.

Expand 1 Items
Loading...
Samco™ Round Base Test Tubes, 12×75 mm, Thermo Scientific

Samco™ Round Base Test Tubes, 12×75 mm, Thermo Scientific

Supplier: Samco

Samco™ round based 12×75 mm tubes are suitable for most laboratory applications.

Expand 5 Items
Loading...

Sample Tube, Ultra Thin-Bore, Thermo Scientific

Supplier: Thermo Scientific Chemicals

Expand 1 Items
Loading...
Transport Tubes, Starplex Scientific

Transport Tubes, Starplex Scientific

Supplier: Starplex

Threadless polypropylene round bottom tubes fit easily into racking or automated systems.

Expand 3 Items
Loading...
Sample Tube, Ace Glass Incorporated

Sample Tube, Ace Glass Incorporated

Supplier: Ace Glass

Designed by Dr. H.R. Krouse, University of Calgary, for use with H₂O¹⁸/H₂O¹⁶ determinations.

Expand 1 Items
Loading...
Pressure Reactor Sparger Tubes, PTFE, Ace Glass Incorporated

Pressure Reactor Sparger Tubes, PTFE, Ace Glass Incorporated

Supplier: Ace Glass

This sparger tube is designed for use in #15 Ace-Thred with either one-piece or two-piece reactors.

Expand 5 Items
Loading...
GC Collection Tube, Ace Glass Incorporated

GC Collection Tube, Ace Glass Incorporated

Supplier: Ace Glass

Microscale gas chromatography collection tube for fraction collection followed by transfer of sample to Ace Glass' conical reaction 0.1 ml vial for storage.

Expand 1 Items
Loading...
Receiving Tube/Cylinder, Ace Glass Incorporated

Receiving Tube/Cylinder, Ace Glass Incorporated

Supplier: Ace Glass

This graduated, tapered bottom, receiving tube is a replacement for receivers on all glassware sets supplied with Heidolph® LR4000 Series rotary evaporators or with other evaporators.

Expand 1 Items
Loading...
SP Wilmad-LabGlass Schlenk Vacuum Sample Tubes, SP Industries

SP Wilmad-LabGlass Schlenk Vacuum Sample Tubes, SP Industries

Supplier: SP Industries

These borosilicate glass Schlenk sample tubes incorporate a grease-free high vacuum 0 to 4 mm valve and are suited for pressure/vacuum applications.

Expand 1 Items
Loading...
Disposable Polypropylene Snap Cap Test Tubes

Disposable Polypropylene Snap Cap Test Tubes

Supplier: Corning

Ensured sterility with the convenience of disposability makes these tubes ideal for all types of laboratory procedures.

Expand 2 Items
Loading...

Screw Thread Culture Tubes, Kimble Chase

Supplier: DWK Life Sciences (KIMBLE)

These borosilicate culture tubes are suitable for tissue culture work and general bacteriological use.

Expand 4 Items
Loading...
KIMBLE® Vacuum Hydrolysis Tubes, DWK Life Sciences

KIMBLE® Vacuum Hydrolysis Tubes, DWK Life Sciences

Supplier: DWK Life Sciences (KIMBLE)

Versatile tube for use where a vacuum environment is desired.

Expand 3 Items
Loading...
Collection Tubes 2 ml, Zymo Research

Collection Tubes 2 ml, Zymo Research

Supplier: Zymo Research

Collection tubes (2 ml) compatible with Zymo-spin line of columns.

Expand 3 Items
Loading...
AIRLESS-WARE® Schlenk Vacuum Tubes, Kimble Chase

AIRLESS-WARE® Schlenk Vacuum Tubes, Kimble Chase

Supplier: DWK Life Sciences (KIMBLE)

This Schlenk tube is suitable for vacuum gas manifolds with HI-VAC® valve to control gas flow to the reaction tube.

Expand 5 Items
Loading...
Storage Tube, Micro Schlenk, Ace Glass Incorporated

Storage Tube, Micro Schlenk, Ace Glass Incorporated

Supplier: Ace Glass

This tube is used for storage or simple reactions.

Expand 2 Items
Loading...
Amber Plain Disposable Borosilicate Glass Tubes, Kimble Chase

Amber Plain Disposable Borosilicate Glass Tubes, Kimble Chase

Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX

Premium quality tubes provide superior protection for light sensitive applications.

Expand 4 Items
Loading...
KIMBLE® Schlenk tubes, DWK Life Sciences

KIMBLE® Schlenk tubes, DWK Life Sciences

Supplier: DWK Life Sciences (KIMBLE)

These Schlenk tubes are designed to minimize the loss of volatile solvents while manipulating under normal conditions.

Expand 2 Items
Loading...
Disposable Borosilicate Glass Tube

Disposable Borosilicate Glass Tube

Supplier: DWK Life Sciences (KIMBLE)

Plain disposable borosilicate glass tube.

Expand 1 Items
Loading...
Nunc® Externally-Threaded Universal Tubes, 1.8 ml, Thermo Scientific

Nunc® Externally-Threaded Universal Tubes, 1.8 ml, Thermo Scientific

Supplier: Thermo Fisher Scientific

Versatile Nunc® 1.8 ml externally-threaded universal tubes can be used inside a large storage system, or in smaller scale applications such as traditional cryostorage in dewars.

Expand 4 Items
Loading...
Nunc® Internally-Threaded Universal Tubes and Rack System, 2.0 ml, Thermo Scientific

Nunc® Internally-Threaded Universal Tubes and Rack System, 2.0 ml, Thermo Scientific

Supplier: Thermo Fisher Scientific

The Nunc® 2.0 ml internally-threaded universal tubes are available racked or in bulk, with non-coded or 2D barcodes to serve every need.

Expand 4 Items
Loading...
Sort by
Recommended for You