You searched for: Chromatography Columns
Chromatography columns separate molecules within mixtures during gas or liquid methods. Filled with stationary phase compositions or resins, the prepacked options reduce the preparing time required. The vertical pieces are able to withstand extreme temperatures and pressure fluctuations experienced when the mobile phase is forced through the medium. Meet preparative and analytical laboratory procedures with the chromatography columns to separate dissimilar moving components based on individual polarity, composition, and size.
Avantor® Hichrom HI-JXR, GC Columns
Supplier: VWR International
General purpose apolar column.
Expand 157 Items
Avantor® Hichrom HI-PS255, GC Columns
Supplier: VWR International
Apolar phase, suitable for high flm thickness columns, to analyze solvents, alcohols, volatiles, and so on.
Expand 170 Items
Avantor® Hichrom HI-1 HT, GC Columns
Supplier: VWR International
General purpose GC-MS column Solvent impurities, PCBs, SimDist, drugs, natural gases, essential oils, semivolatiles, pesticides, phenols, etc.
Expand 24 Items
Avantor® Hichrom HI-DEX DAC Beta, GC Columns
Supplier: VWR International
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Expand 11 Items
Avantor® Hichrom HI-DEX DET Beta, GC Columns
Supplier: VWR International
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Expand 11 Items
Avantor® Hichrom HI-1701, GC Columns
Supplier: VWR International
General purpose low-to-mid-polarity phase for a wide range of applications, including Aroclors, pesticides, herbicides and more.
Expand 139 Items
Avantor® Hichrom HI-624 MS, GC Columns
Supplier: VWR International
General purpose low-to-mid-polarity phase for a wide range of applications, including (for example): Residual solvents, volatile pollutants. HI-624 MS is an MS grade crossbonded column.
Expand 150 Items
Avantor® Hichrom UNCORET, GC Columns
Supplier: VWR International
Large volume injection pre-columns.
Expand 2 Items
Avantor® Hichrom HI-35 MS, GC Columns
Supplier: VWR International
Proprietary low-polarity and low-bleeding stationary phase; great choice for pesticides, herbicides, polychlorinated biphenyls (PCBs), and polyaromatic hydrocarbons (PAHs). Ideal for GC/MS applications and is also an excellent choice as a confirmation column for semivolatile environmental analysis.
Expand 149 Items
Avantor® Hichrom HI-17 MS, GC Columns
Supplier: VWR International
Really versatile general purpose polar phase for a wide range of applications. Ideal in confirmation analysis. HI-17 MS is an MS grade low-bleeding crossbonded column and assure a long lifetime even under high temperature limits.
Expand 148 Items
Avantor® Hichrom HI-TCEP, GC Columns
Supplier: VWR International
Application specifc columns ideal for aromatics and oxygenates in gasoline.
Expand 4 Items
Avantor® Hichrom HI-LAP, GC Columns
Supplier: VWR International
Application specific column for lipids, sterols and triglycerides analysis.
Expand 1 Items
Avantor® Hichrom HI-PLUS, GC Columns
Supplier: VWR International
Offering a range of selectivities with differing copolymer rations of PEG and methyl polysiloxane. HI-PLUS, HI-PLUS 10, HI-PLUS 25, HI-PLUS 75, HI-PLUS 90.
Expand 123 Items
Avantor® Hichrom HI-65 HT, GC Columns
Supplier: VWR International
High Temperature column ideal for triglycerides separations based on carbon number and degree of unsaturation.
Expand 5 Items
Avantor® Hichrom HI-BIODIESEL 105, GC Columns
Supplier: VWR International
Application specific column for free and total glycerine (phase stable up to 380 °C) and for FAMEs in biodiesel analysis.
Expand 3 Items
Avantor® Hichrom HI-5 MS, GC Columns
Supplier: VWR International
Really versatile general purpose low-polarity phase for a wide range of applications, including (for example): PCBs, essential oils, pesticides, semivolatiles and many more. HI-5 MS is an MS grade low-bleeding crossbonded column and assure a long lifetime even under high temperature limits.
Expand 148 Items
Avantor® Hichrom HI-1701 HT, GC Columns
Supplier: VWR International
High temperature general purpose column. Ideal for confrmation analysis, ideal as mid-polar column in GC×GC-HT confgurations.
Expand 23 Items
Avantor® Hichrom HI-PS264, GC Columns
Supplier: VWR International
Apolar phase, suitable for high flm thickness columns, to analyze solvents, alcohols, volatiles, and so on.
Expand 171 Items
Avantor® Hichrom HI-5 MS Xil, GC Columns
Supplier: VWR International
Silphenylene based stationary phase (selectivity equivalent to 5MS columns) to ensure minimum bleeding level at high temperatures. General purpose low-polarity phase for a wide range of applications, including (for example): PCBs, essential oils, pesticides, semivolatiles and many more. HI-5 MS Xil is an MS grade low-bleeding crossbonded column and assure a long lifetime even under high temperature limits.
Expand 148 Items
Avantor® Hichrom HI-200, GC Columns
Supplier: VWR International
Unique selectivity trifluoropropyl methyl polysiloxane stationary phase.
Expand 126 Items
Avantor® Hichrom HI-5 Basic, GC Columns
Supplier: VWR International
Max temperature limits may change depending on the film thickness.
Expand 24 Items
Avantor® Hichrom HI-SE52, GC Columns
Supplier: VWR International
General purpose column for solvent impurities, PCBs, hydrocarbons, essential oils, semivolatiles, triglycerides, pesticides, poly-waxes, and so on.
Expand 165 Items
Avantor® Hichrom HI-DEX DMP Beta, GC Columns
Supplier: VWR International
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Expand 10 Items
Avantor® Hichrom HI-SE54, GC Columns
Supplier: VWR International
General purpose column for solvent impurities, PCBs, hydrocarbons, essential oils, semivolatiles, allergens, pesticides, and so on.
Expand 165 Items
Avantor® Hichrom HI-WAX HT, GC Columns
Supplier: VWR International
High Temperature unique PEG phase. Extend the temperature limits of your FAST-GC and GCxGC methods while using a polar WAX phase.
Expand 23 Items
Avantor® Hichrom HI-1301, GC Columns
Supplier: VWR International
Max temperature limits may change depending on the film thickness.
Expand 146 Items
Avantor® Hichrom HI-XMLB, GC Columns
Supplier: VWR International
Proprietary low-polarity and low-bleeding stationary phase; great choice for pesticides, herbicides, polychlorinated biphenyls (PCBs), and polyaromatic hydrocarbons (PAHs). Ideal for GC/MS applications and is also an excellent choice as a confirmation column for semivolatile environmental analysis.
Expand 18 Items
Avantor® Hichrom HI-PAH 2, GC Columns
Supplier: VWR International
Application specifc column for polycyclic aromatics hydrocarbons.
Expand 4 Items
Avantor® Hichrom HI-ALC 1, GC Columns
Supplier: VWR International
Application specific column for blood alcohols testing.
Expand 3 Items
Avantor® Hichrom HI-ALC 2, GC Columns
Supplier: VWR International
Application specific column for blood alcohols testing.