Orders LinkContactUsLinkComponent
Block_1_sf_1817.jpg
SureSTART Autosampler Vials and Wellplates
3 distinct performance levels-differentiated by performance and value-make selecting the right Thermo Scientific SureSTART product simple, clear and easy. 
SureSTART Autosampler Vials and Wellplates | Avantor
Power to extend your consumables budget 
Realize extra value with built-in bulk discounts - the more you buy, the more you save.
Assured quality, performance, and compatibility
Suitable for all current HPLC/UHPLC or GC instrument brands. With over 40 years of experience supplying vial and well plates, SureSTART certified products meet ‘critical to quality’ specifications, consistently. 
We’ve got you covered - from incorporating our innovative patented products that minimize sample evaporation, to the curated selection ensuring sample security and low compound adsorption, you can meet your regulatory requirements with ease.
Explore SureSTART™ Sample Handling Solutions
Ad1_sf_1841.png
Delivering a new level of performance, reproducibility and accuracy of results.
Ad3_sf_1841.jpg
Suitable for QA/QC applications using core detectors, non-MS.:UV/Vis, PDA, RI, ELSD, FLD, FID, NPD, ECD, TCD, or FPD.
Ad4_sf_1841.jpg
Suitable for routine QA/QC applications using MS tech. Compound concentrations are at easily detectable or low levels of detection.
Ad5_sf_1841.png
Suitable for challenging and research-focused applications, using MS tech. Compound concentrations are at/near the method detection limit.