Orders LinkContactUsLinkComponent
protein-therapy-955.jpg
Protein Therapy Chromatography Products
Innovative, reliable and easy-to-use, Thermo Scientific™ solutions help you meet the demanding needs at every stage of your protein therapy workflow. From sample preparation to sample security and separation, we have the most comprehensive range of chromatography consumables.
Protein Therapy Chromatography Products | Avantor
Product Selection
Hypersil-GOLD-400.jpg
Offers chromatographers outstanding peak shape for reversed phase, ion exchange, HILIC or normal phase chromatography.
GOLD-Grade-400.jpg
Best for analyzing analytes such as polar compounds that strongly absorb on the glass surface or are sensitive to sample pH changes.
MAbPac-495.jpg
For high-resolution separation of monoclonal antibody (mAb) analysis, including the monomers, aggregates and Fab and Fc fragments resulting from proteolysis.
ProPac-495.jpg
Achieve ultrahigh resolution and high efficiency separations of proteins and glycoproteins, monoclonal antibodies, biosimilars, and charge variants.
SMAR-digest-kits.jpg
Highly stable immobilized trypsin reactor; when combined with state-of-the-art heating technology, it produces fast, reproducible digestions ready for analysis by LC/MS.
Accucore_400.jpg
Combined with the larger pore size of the solid core particles, makes it well suited for separating a carbohydrates and peptides.
Polypropylene-400.jpg
Ultimate performance and sensitivity for your methods, while maintaining the highest levels of sample security and integrity. Polypropylene caps designed to reduce autosampler needle issues.
HIC-20-400.jpg
Used for separation of monoclonal antibodies (mAbs), mAb fragments and oxidized variants by hydrophobic interaction chromatography (HIC). The unique chemistry of this silica column offers high resolution and excellent selectivity.
HIC-20-400.jpg
Use to separate monoclonal antibodies (mAbs) and antibody-drug conjugates (ADCs). This column is based on hydrophilic, nonporous, 5μm polymer particles functionalized with butyl groups.