Orders LinkContactUsLinkComponent
Hero1_sf_1818.jpg
Pharmaceutical Chromatography Products
Innovative, reliable and easy-to-use, Thermo Scientific™ solutions help you meet the demanding needs at every stage of your pharmaceutical workflow. From sample preparation to sample security and separation, we have the most comprehensive range of chromatography consumables.
Chromatography Consumables for Pharmaceutical Workflows | Avantor
Product selection
Block 1 _sf_1818.jpg
HPLC columns which are used to achieve consistent, reproducible separations, ideal for the demands of QA/QC laboratories.
sure-stop-vials.jpg
Remove subjectivity around achieving optimal compression when sealing a vial. Improve analytical results and reduce analysis error rate that results from the evapouration of solvent in improperly sealed vials.
Hypersil-GOLD-400.jpg
Offers chromatographers outstanding peak shape for reversed phase, ion exchange, HILIC or normal phase chromatography.
Solid-Phase-400.jpg
Provides unparalleled performance characteristics for bio-analysis compared to conventional SPIE due to the innovative fritless design.
Block 5_sf_1818.jpg
Solid core particles enabling high speed, high resolution separations with back pressures significantly lower than those associated with UHPLC.
web-seal.jpg
A comprehensive range of well plate solutions enabling high sample integrity for high density sample handling in chromatography.