Orders LinkContactUsLinkComponent
Silver-Products-Feature.jpg
Specialty Silver Products
Silver products are used extensively in a wide range of applications from medical & pharmaceutical to catalysis, energy & advanced optoelectronics that touch everyday life.
Specialty Silver Products | Avantor
undefined
Literature
Some of the key features of our silver products include: silver compounds are manufactured up to a purity of 99.999%, a batch specific certificate of analysis is produced for each product, each silver product batch has full metallic impurities typically measured by ICP-MS to ppm and measured silver content.
Product selection