Orders LinkContactUsLinkComponent
IndicatorsWF.png
Chemical Indicators
RICCA offers a wide variety of Chemical Indicators suitable for most titration applications. Options include Color Indicators covering the 0-13 pH range as well as other reaction types. RICCA's manufacturing processes ensure the sharpest color changes and the crispest endpoints, giving you the most accurate results. Test strips are also available for rapid pH analysis!
Chemical-Indicators | Avantor
Broad Range of Indicators for:
  • Adsorption Titrations
  • Complexometric Titrations
  • Oxidation-Reduction Titrations
  • Acid-Base Titrations
  • Argentometric Titrations
Product selection
PhenolphthaleinWF.jpg
Suitable for acid-base titrations in aqueous and non-aqueous applications. Get peak titration performance with RICCA.
Potassium-ChromateWF.jpg
For use in argentometric titrations using silver nitrate. Enhance your analysis with RICCA's potassium chromate indicators.
Starch.jpg
Great for use in redox titrations of iodine! Crisp color changes that increase your accuracy and precision.
Test-StripsWF.jpg
pH Test Strips
Need rapid pH testing? Look no further, RICCA has test strips for quick readings across a variety of pH ranges.