ToughMix® qPCR MasterMixes
PCR & qPCR ToughMixes Engineered For Difficult Samples
- Overcome PCR inhibition
- Robust fluorescent probe qPCR & PCR performance
- Superior performance with pure template
- Applicable to wider range of probe designs and detection chemistries
Inhibitors | Common Source |
Polysaccharides | Plants, seeds, stool |
Heme, hemoglobin | Blood |
Humic acid | Soil, plant material |
Melanin | Skin, hair |
Product selection
Highly sensitive, ready-to-use master mix for RT-qPCR of RNA templates using probe-based detection.
4x master mix for highly sensitive 1-step RNA detection using multiplexed probe-based RT-qPCR with crude extracts or inhibitory biological starting materials.
5x ready-to-use master mix that supports highly multiplexed qPCR across a range of inhibitory sample types and crude extraction methods.
2x master mix for accurate SNP genotyping with crude extracts, clinical specimens, environmental samples, plant/animal tissues, or complex food matrices.