Orders LinkContactUsLinkComponent
puritan-feature.jpg
Swabs for Testing Coronavirus (COVID-19)
When conducting diagnostic testing for COVID-19, CDC recommends Nasopharyngeal (NP) or Oropharyngeal (OP) swabbing. Puritan's foam or flock swabs are the ideal swab for nasopharyngeal and oropharyngeal COVID-19 specimen collection.
Video
Puritan - Swabs for Testing Coronavirus (COVID-19) | Avantor
COVID-19 testing is continually evolving. When conducting diagnostic testing for COVID-19, the CDC recommends Nasopharyngeal (NP) or Oropharyngeal (OP) swabbing. Depending on how the platform used to test the specimen, some tests are designed to analyze a swab specimen directly, without the use of any media.
Other platforms require the swabs to be placed immediately into a sterile transport tube containing 2-3mL of either UniTranz-RT Universal Transport Systems, Opti-Swab® - Liquid Amies Transport Systems, or sterile saline solution. Our foam or flock swabs are the ideal swab for nasopharyngeal and oropharyngeal specimen collection when 2019-nCoV is suspected.
hydra_flock.jpg
HydraFlock provides superior collection and a quick release of specimens, enhanced by its unique microstructure with multi-length fibers. PurFlock Ultra’s high purity features make it an ideal swab for rapid diagnostic tests.
foam-swab.jpg
This specialty applicator is tipped with 100ppi medical grade polyurethane foam. Ideal for nasal, anterior nares, buccal specimen collection for testing and screening.
UniTranz-RT.jpg
Puritan UniTranz-RT™ Collection and Transport System is intended for the collection and transport of clinical samples containing viruses, chlamydiae, mycoplasmas and ureaplasmas from the collection site to the testing laboratory...
Opti-Swab.jpg
The Puritan Opti-Swab Liquid Amies Collection and Transport System consists of a sterile peel-open pouch containing a pre-scored HydraFlock® flocked swab and a polypropylene screw-cap vial containing 1 ml of modified liquid Amies medium.