ContactUsLinkComponent
polyplus955x590.jpg
mRNA transfection reagent - jetMESSENGER
jetMESSENGER is a mRNA transfection reagent that has been specifically designed to offer outstanding transfection efficiency in cells that are usually hard to transfect, allowing relevant gene expression experiments in all cell types.
  • Unmatched transfection efficiency
  • High efficiency on a wide variety of difficult to transfect cells
  • Outperform DNA transfection by switching to mRNA
  • Extremely gentle on cells
  • No risk of genome integration
  • Perfectly suited for CRISPR/Cas9 gene editing, iPS generation, stem cell differentiation and immunotherapy assays
Request your free sample today
Description
jetMESSENGER has been designed to transfect cells that are slow dividing or that have developed specific mechanisms such as primary cells, neurons, suspension cells and various cancer cell lines. jetMESSENGER is a highly efficient and gentle mRNA transfection reagent. mRNA transfection presents many advantages versus DNA transfection:
  • no risk of genome integration, hence no genome modification of the transfected cell
  • no promoter regulation issue
  • no need to reach the nucleus for efficient expression
  • more gentle process
jetMESSENGER has been specifically designed to offer outstanding transfection efficiency in cells that are usually hard to transfect, finally allowing relevant gene expression experiments in almost all cell types.
chart1920x760.jpg
jetMESSENGER outperforms its main DNA transfection reagent competitor. Transfection efficiency was assessed by FACS analysis in various cell lines 24 h after transfection of eGFP mRNA (5meC, pseudo-uridine, Trilink) or plasmid DNA encoding for eGFP. Conditions were used according to the manufacturers’ recommendation.
With jetMESSENGER, achieve outstanding transfection efficiency in a wide variety of cell lines, from adherent and suspension cancer cells, to sensitive primary cells and neurons. jetMESSENGER is also perfectly suitable for reprogramming experiments in fibroblasts and stem cells.
mRNA-table1920x760.jpg
jetMESSENGER leads to high transfection efficiency in a wide variety of cell types.
By eliminating the need to reach the nucleus for efficient expression, jetMESSENGER allows transfection of quiescent and slow dividing cells. In addition, jetMESSENGER operates through an extremely gentle process. Cell viability remains extremely high during transfection and cell morphology is maintained. Make your transfection experiments physiologically relevant!
cell-types-l-1920x760.jpg
jetMESSENGER provides a better cell viability and a higher protein expression than DNA transfection. Primary Rat Cortical neurons, Hep G2 and mouse stem cells were analyzed 48 h after transfection using phase contrast and fluorescent microscopy. The transfections were performed with jetMESSENGER/eGFP mRNA (5meC, pseudo-uridine, Trilink) and L2K/eGFP plasmid DNA according to the manufacturers’ recommendations for each reagent.
Examples of citations
Viegas C.S.B., Costa R.M., Santos L., Videira P.A., Silva Z., Araújo N., Macedo A.L., Matos AP., Vermeer C., Simes D.C. (2017). Gla-rich protein function as an anti-inflammatory agent in monocytes/macrophages: Implications for calcification-related chronic inflammatory diseases. PLoS Ones, DOI:10.1371/journal.pone.0177829
Sultana N., Magadum A., Hadas Y., Kondrat J., Singh N., Youssef E., Calderon D., Chepurko E., Dubois N., Hajjar RJ., Zangi L. (2017). Optimizing Cardiac Delivery of Modified mRNA. Mol Ther, DOI:10.1016/j.ymthe.2017.03.016
Testimonials
"Application of jetMESSENGER reagent to introduce modified RNA molecules into primary lung mouse endothelial cells (a cell population which is generally hard to transfect with DNA/RNA using non-viral methodology) resulted with the highest transfection rate (>70%) and with the lowest toxicity effects as compared to other tested RNA/DNA transfection reagents (jetPEI, TransIT, & RNAiMAX)."
Itkin T., Weill Cornell Medicine, New York, United States
Product selection
polyplus540x390.jpg
jetPRIME® DNA transfection reagent
Do you perform transient gene expression? jetPRIME is a powerful and versatile DNA and siRNA transfection reagent for day-to-day experiments that leads to efficient and reliable scientific results.
vivo-jetPEI540x390.jpg
in vivo-jetPEI® for in vivo nucleic acid delivery
in vivo-jetPEI is a powerful polymer based reagent used to deliver any nucleic acid to any animal model. in vivo-jetPEI has already been used to target a wide range of organs using various administration routes.
jetCRISPR540x390.jpg
jetCRISPR is an innovative reagent especially designed to deliver RNP in a CRISPR/Cas9 experiment. Use the leading technology for CRISPR/Cas9 RNP delivery!