Orders LinkContactUsLinkComponent
4677896-feature.jpg
Chromatography
Innovative Solutions for all Applications Since the early days of chromatography, MilliporeSigma has introduced numerous products that have been recognized as milestones in the development of chromatography technology. This spirit of innovation still continues relentlessly.
Chromatography | Avantor
Today, MilliporeSigma is among the leaders in the science of liquid chromatography and is committed to the further development of sorbents, analytical and preparative columns, sample preparation, and thin layer chromatography products.
Related Information:
AT-HPLC-EMD-041515-V01_MP4_VWR_260x190.jpg
MilliporeSigma HPLC Innovations
EMDM_TLC_100.jpg
Thin-Layer Chromatograhy (TLC and HPTLC)
As a pioneer in TLC, EMD Millipore introduced the first pre-coated plates on the market. Today, we offer a wide range of reliable plates and regularly roll out new products.
EMDM-analytical-HPLC-100.jpg
Analytical HPLC
With Chromolith® columns you are able to perform fast and robust separations using standard HPLC systems, thanks to the unique, patented, monolithic silica technology. For all polar and hydrophilic compounds, the proprietary zwitterionic SeQuant® ZIC®-HILIC technology gives you straightforward HPLC separations with high flexibility in the selection of separation conditions. The optimally balanced selectivity of Purospher® makes it the perfect choice for reversed phase method development in a wide variety of labs. MilliporeSigma’s renowned HPLC column brands, LiChrosorb®, LiChrospher® and Superspher® continue to provide the results you expect.
EMDM-gas-chromatography-100.jpg
Gas Chromatography
A wide range of high-purity solvents are available, optimized for highly sensitive applications, such as pesticide residue and trace analyses.
EMDM-mobile-phase-100.jpg
Mobile Phases for Chromatography
High-Purity Solvents for Reliable, Reproducible Results