Orders LinkContactUsLinkComponent
Skin and Glove Cleansing and Sanitation | Avantor
Skin and Glove Cleansing and Sanitation
Micronova's Hand Hygiene products are an important part of keeping employees and products clean and safe from debris, microbials, and viruses. Products range from cleansing wipes to hand soaps and glove sanitizers which can be used with VWR® MicroDispenser™ Touch-Free Soap Dispenser.
NovaClenz_Micronova_200.jpg
NovaClenz™ Instant Hand Sanitizer
Highly effective, fast-acting instant hand sanitizer ideal for use under gloves. Contains 65% minimum ethanol which helps to prevent viral transfer. Requires no sink, water, or towels. Effective against bacteria, including: pseudonomas, salmonella, and staphylococcus and reducing nosocomial infections.
AquaHol_Micronova_200.jpg
AquaHol™ Sterile Isopropyl Alcohol Glove Sanitizer for Touch-Free Dispenser
Sterile hand and glove sanitizer comprised of 70% isopropyl alcohol with 30% WFI quality purified water. Ideal for use over gloves prior to entering into aseptic areas. Meets USP requirements. Terminally filtered to 0.1 microns and gamma irradiated. Certificate of Sterility.
Hygclenz_Micronova_200.jpg
Hy-G-Clenz™ Antiseptic Hand Soap
Anti-microbial soap made with 0.13% Benzalkonium Chloride and natural emollients. Kills germs and leaves hands feeling soft and smooth. Persistent activity for up to three hours. Ideal for frequent hand washing.
Novadyne_Micronova_200.jpg
NovaDyne™ pH Balanced General Purpose Lotion Hand Soap
Gentle yet effective general purpose hand soap for use in office and general industrial spaces.
AboutFace_CleansingWipes_Micronova_200.jpg
Individually packaged poly-blend cleansing wipe that is pre-moistened with a gentle, alcohol-free cleansing lotion, designed for the face and hands. Highly effective as a make-up remover; ideal for pre-gown rooms.