Orders LinkContactUsLinkComponent
XPert-Feature.jpg
XPert® Balance Systems & Enclosures
The XPert® line of balance and bulk powder enclosures provide user protection by containing airborne powders and particulates.
XPert® Balance Systems & Enclosures | Avantor
Labconco's™ industry leading XPert Balance Systems and Enclosures provide user protection by keeping powders and particulates contained during manipulations such as weighing. The System features a built-in blower and exhaust HEPA filter and takes up less space on the bench. Vibration isolation allows for steady weighing with a microbalance. Enclosure models are flexible and may be hooked up to a portable exhauster or filtered house exhaust.
Now through the REDISHIP program, Labconco XPert Systems and Enclosures are available within 48 hours.
Certifications: Independently validated by SafeBridge Consultants. Tested to Modified ASHRAE 110. ETL listed. Contact Avantor to see how you can maximize protection from hazardous powders while minimizing space requirements.
XPert-Balance-Systems.jpg
XPert® Balance Systems
Balance Systems provide maximum user protection by keeping powders and particulates contained during weighing operations. Trusted by major global pharmaceutical organizations, XPert Systems keep operators safe from potent active pharmaceutical ingredients (APIs).
XPert-Balance-Enclosures-Feature.jpg
XPert® Balance Enclosures
Enclosures are flexible powder containment hoods that rely on external exhaust or local filtered exhaust systems. Available in sizes up to 8' wide for all types of powder handling processes. Storage cabinet sold separately.
Bag-Column.jpg
Bag-In/Bag-Out Safe HEPA Filter System
Allows for safe exchange of the HEPA filter without contaminating the surrounding laboratory area.
built-to-last.jpg
Built To Last
Tempered safety glass and metal construction offer excellent visibility, ambient light, and long-term durability.