Orders LinkContactUsLinkComponent
Halo_Biopharma955-590.png
HALO® HPLC Columns for Biopharma Separations
Advancing the understanding of form and function for biomolecules, researchers supporting their development and release employ many orthogonal techniques and separation methodologies to develop a more complete therapeutic picture.
    Biopharma Columns Literature
    HALO HPLC Columns for Biopharma Separations | Avantor
    With both tailored particle and pore size options, HALO® BIOCLASS offers application-specific solutions for:
    • Intact proteins, monoclonal antibodies (mAbs), biosimilars and other large biomolecules such as pegylated proteins, antibody-drug conjugates (ADCs), etc.
    • Peptide mapping (analysis of enzyme digests) for characterization and monitoring of synthetic protein drugs
    • Analysis of therapeutic peptides and peptide biomarkers (protein surrogates)
    • High-resolution separations of complex mixtures of glycans released from N- and O-linked glycoproteins
    PROTEIN SEPARATIONS
    OLIGIONUCLEOTIDES
    PEPTIDE SEPARATIONS
    GLYCAN SEPARATIONS