Orders LinkContactUsLinkComponent
Dignostic-955x690.jpg
Diagnostic
In your quest to deliver consistent, reliable, and reproducible results, you need a partner you can count on to supply you with a broad range of laboratory and diagnostic consumables and equipment.
Corning Diagnostic | Avantor
To deliver them when and where you need them. To provide expert service and support. And to understand your needs as you work towards a laboratory developed test (LDT) or test execution. Corning’s high quality lab consumables and equipment work across your workflow of collection, sample prep, and assay to help you meet your objectives.
Corning® Lambda™ EliteTouch™ Starter Kit, Corning
CorningLambdaEliteTouchStarterKit-540x390.jpg
The Lambda EliteTouch Starter Kit is designed for laboratories which are about to replace an old set of pipettors, and for customers who want to get a new set of pipettors which cover the most commonly used volumes.
  • Three-year manufacturer warranty included
Certifications: Calibration certificate under ISO 8655.
Ordering information: Starter Kit includes: 4 single-channel pipettors: 0.5 to 10 µl (item 6051), 2 to 20 µl (item 6052), 20 to 200 µl (item 6055), 100 to 1000 µl (item 6056), universal linear stand for four single-channel pipettors (item 6066), Corning® DeckWorks pipet tips: 10 µl (item 4135), 200 µl (item 4138), 1000 µl (item 4140), 3 colored pushbuttons (4 sets).
Order here
Literature