Orders LinkContactUsLinkComponent
Biotix Plates | Avantor
Biotix Plates
Biotix Assay and Deep Well Plates are designed for high throughput screening; engineered with raised alpha/numeric grids for clear well identification, sample tracking and plate orientation. All plates are manufactured with non-treated, medical-grade virgin polypropylene making them biologically inert with great temperature and chemical resistance.