Orders LinkContactUsLinkComponent
CytoFLEX-Linear-Layout_774.jpg
CytoFLEX: Benchtop Flow Cytometry Without Compromises
Beckman Coulter CytoFLEX | Avantor
The CytoFLEX Flow Cytometer provides the performance you need in an easy to use system allowing you to focus on the science, not the instrumentation. The CytoFLEX Flow Cytometer’s superior sensitivity and resolution throughout all configurations give it the edge over other cytometry systems four times its size.
Leverage the power of the CytoFLEX:
  • Exquisite sensitivity
  • Nanoparticles analysis in a benchtop analyzer
  • Extensive set of repositionable band pass filters
  • Flexibility to upgrade by adding additional parameters
  • Intuitive software to facilitate multicolor analysis
Request a Demo
Request a Quote
CytoFLEX.jpg
The CytoFLEX has a unique flow cell design with Integrated Optics as well as an innovative Wavelength Division Multiplexing detection module.
View video
CytoFLEX.jpg
The high-performance CytoFLEX system offers scalable detection capabilities for up-to 13-color research flow cytometry right on your benchtop.
CytExpert-Software-for-CytoFLEX.jpg
Cytexpert acquisition and analysis software has an intuitive workflow capabilities for complex flow cytometric applications.
undefined
CytoFLEX Application Notes
Download the application notes Flipbook to learn about different applications that can be performed on the CytoFLEX system.
Download PDF