Orders LinkContactUsLinkComponent
AzureAdvancedImagingSystems955x590.jpg
Azure Advanced Imaging Systems
The Azure 300-600 offer imaging capabilities for chemiluminescence, UV, RGB, NIR, blue light, and white light, depending on the model that fits your needs. The 600 offers the most flexible imaging options, but each model can be upgraded to suit your lab’s needs as they change.
Azure Advanced Imaging Systems | Avantor