Orders LinkContactUsLinkComponent
Norm-Ject.gif
HSW® Norm-Ject® syringes
Latex-free and containing no rubber, silicone oil, styrene, BPA, or DEHP, Henke-Sass Wolf’s (HSW) Norm-Ject syringes are the choice for any situation needing inert, hypo-allergenic syringes.
HSW® Norm-Ject® syringes | Avantor
Due to their purity, Norm-Ject are indicated for in vitro fertilization (IVF), embryo-transfer, chromatography, and many laboratory procedures. Manufactured only from laboratory-grade polypropylene and polyethylene, these unique plastic syringes have a positive safety stop to prevent accidental spills. Available in sterile individual packs or in bulk, unsterile packaging.
Product selection
NormJect_Luerslip.jpg
The all-plastic syringe with a Luer slip.
NormJec_-Luerlock.jpg
The all-plastic syringe with a Luer lock.
NormJect_bulkSyringes.jpg
Norm-Ject bulk syringes
Unsterile Norm-Ject syringes in bulk.