Order Entry
ContactUsLinkComponent
12594 results for Proteins and Peptides

You searched for: Proteins and Peptides

Proteins and Peptides

Proteins are used in routine laboratory procedures such as binding enzymes or coupling peptides to carrier proteins. These kits, mixture solutions, and collagen matrices fulfill a myriad of essential laboratory functions for developing relationships between proteins and other cellular components. The stimulating proteins offered have various amino acid arrangements and functions to fulfill any sample manipulation for testing purposes in any field.

Sort by
Gelatin

Gelatin

Supplier: VWR

Gelatin

Expand 2 Items
Loading...
Bovine Serum Albumin (BSA)
Expand 2 Items
Loading...
Aprotinin

Aprotinin

Supplier: VWR

Product is animal-free

Expand 1 Items
Loading...
VWR Life Science Ribonuclease Inhibitor (RNase Inhibitor)

VWR Life Science Ribonuclease Inhibitor (RNase Inhibitor)

Supplier: VWR

RNase Inhibitor is used to inhibit the activity of RNases in reaction mixtures for cDNA synthesis and in vitro transcription, as well as for long term storage of valuable RNA samples

Expand 1 Items
Loading...
Aprotinin

Aprotinin

Supplier: VWR

A competitive and reversible serine protease inhibitor of chymotrypsin, trypsin, kallikrein and plasmin

Expand 1 Items
Loading...
Leupeptin

Leupeptin

Supplier: VWR

Product is animal-free

Expand 1 Items
Loading...
Streptavidin

Streptavidin

Supplier: VWR

A 60 kDa tetrameric protein that exhibits high binding affinity for biotin. Able to bind one molecule of biotin with each subunit, streptavidin is useful in affinity chromatography, ELISA, immunohistochemistry and Western Blotting.

Expand 2 Items
Loading...
Bovine Serum Albumin (BSA)

Bovine Serum Albumin (BSA)

Supplier: VWR

Contains 0.1% sodium azide as preservative and strictly tested for RNase, DNase and Protease making it suitable for sensitive applications.

Expand 2 Items
Loading...
Bovine Serum Albumin (BSA)

Bovine Serum Albumin (BSA)

Supplier: VWR

Bovine serum albumin, powder, Fraction V. Nuclease and protease-free. Suitable for use in most molecular biology applications.

Expand 2 Items
Loading...
Bovine Serum Albumin (BSA)

Bovine Serum Albumin (BSA)

Supplier: VWR

Concentrated BSA solution (Albumin, Bovine, Fraction V – Cold Alcohol Isolation) in 0.85% sodium chloride and contains 0.1% sodium azide (as a preservative for stability).

Expand 1 Items
Loading...
Leupeptin

Leupeptin

Supplier: VWR

A reversible inhibitor of cysteine proteases and serine proteases resembling trypsin

Expand 2 Items
Loading...
Casein

Casein

Supplier: VWR

High purity grade milk protein powder

Expand 1 Items
Loading...
Bovine Serum Albumin (BSA)

Bovine Serum Albumin (BSA)

Supplier: VWR

Crystalline. Prepared by a cold alcohol isolation procedure. This salt-free preparation is the purest material ideal for sensitive biochemical and diagnostic assays. DNase- and RNase-free, virtually free of globulins and other interfering contaminants.

Expand 1 Items
Loading...
J.T.Baker®, Chicken Native Albumin (from Egg)

J.T.Baker®, Chicken Native Albumin (from Egg)

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

CAS RN 9006-50-2. Powder, Purified. Packaged in a plastic container. 500g

Expand 2 Items
Loading...
R18 peptide

R18 peptide

Supplier: Enzo Life Sciences

14-3-3 inhibitor

Expand 1 Items
Loading...
SAMS peptide,Biotin

SAMS peptide,Biotin

Supplier: Enzo Life Sciences

AMPK substrate

Expand 1 Items
Loading...
Angiotensin III

Angiotensin III

Supplier: Enzo Life Sciences

Expand 1 Items
Loading...
Protein Tyrosine Phosphatase Substrate

Protein Tyrosine Phosphatase Substrate

Supplier: Enzo Life Sciences

This peptide sequence, ENDpYINASL, is derived from a highly conserved region of the T-cell phosphatase, TCPTP, and has been shown to be an excellent general substrate for protein tyrosine phosphatases.

Expand 1 Items
Loading...
Ribosomal protein S6 (rat) (229-239), Enzo Life Sciences

Ribosomal protein S6 (rat) (229-239), Enzo Life Sciences

Supplier: Enzo Life Sciences

S6 kinase substrate

Expand 1 Items
Loading...
Human Galanin

Human Galanin

Supplier: Enzo Life Sciences

Neuropeptide

Expand 1 Items
Loading...
Melittin

Melittin

Supplier: Enzo Life Sciences

PLA2 activator

Expand 2 Items
Loading...
E-64

E-64

Supplier: Enzo Life Sciences

Protease inhibitor

Expand 2 Items
Loading...
Spantide III

Spantide III

Supplier: Enzo Life Sciences

Substance P antagonist

Expand 1 Items
Loading...
AC-VAD-AFC

AC-VAD-AFC

Supplier: Enzo Life Sciences

Caspase substrate

Expand 1 Items
Loading...
Z-Aad-Cmk

Z-Aad-Cmk

Supplier: Enzo Life Sciences

Caspase inhibitor

Expand 1 Items
Loading...
Ac-VDVAD-AMC

Ac-VDVAD-AMC

Supplier: Enzo Life Sciences

Caspase-2 inhibitor

Expand 2 Items
Loading...
AC-WEHD-AFC

AC-WEHD-AFC

Supplier: Enzo Life Sciences

Substrate for caspase-1, -4- and -5

Expand 2 Items
Loading...
Human Recombinant ERK1 (from E. coli)

Human Recombinant ERK1 (from E. coli)

Supplier: Enzo Life Sciences

Produced in E. coli.

Expand 1 Items
Loading...
Recombinant DnaK (from E. coli)

Recombinant DnaK (from E. coli)

Supplier: Enzo Life Sciences

Produced in E. coli.

Expand 4 Items
Loading...
Human Recombinant HSP90alpha (from E. coli)

Human Recombinant HSP90alpha (from E. coli)

Supplier: Enzo Life Sciences

Produced in E. coli.

Expand 2 Items
Loading...
Sort by
Recommended for You