Orders LinkContactUsLinkComponent
AVITI-DNA-color.jpg
AVITI™ by Element: Next-generation DNA Sequencing Instrument
AVITI™ leads the way for next-generation sequencing (NGS) to bring affordable, world-class sequencing capability to all labs. Discover how to save resources and improve turnaround times for any sequencing application, at any scale, with a complete range of sequencing kits.
TALK TO AN EXPERT
AVITI™ by Element: Next-generation DNA Sequencing Instrument | Avantor
Features & benefits
Discover the AVITI™ system from Element Biosciences. These benchtop DNA sequencing instruments offer exceptional accuracy and flexible throughput at industry-low sequencing costs and with dedicated scientist-to-scientist support.
Choosing a sequencer doesn’t have to require a trade-off between cost, quality and flexibility:
  • Scalable throughput from 2 billion to 250 million reads per run to suit virtually any application with no tradeoff between TAT and affordability
  • Dual, fully independent flow cells, effectively providing two sequencers in one, each featuring two individually addressable lanes for maximum run flexibility
  • Unprecedented quality, with accuracy ≥70% Q50 and ≥90% Q40 data with Cloudbreak UltraQ™
  • End-to-end workflows from library prep to analysis solutions
The AVITI system includes dedicated support, ensuring smooth installation and expert, scientist-to-scientist assistance.
Shop Related Products
CloudbreakFS-400.jpg
Cloudbreak Freestyle™ builds on the innovation of avidite base chemistry (ABC™) and Cloudbreak™ technology. By enabling linear library loading and circularization on the AVITI™ system, it significantly reduces prep time.
ADD TO CART
CB_Ultra_400.jpg
The Cloudbreak UltraQ™ sequencing reagents deliver the highest accuracy specification on the market today. With multiple strategies to minimize common library prep and sequencing errors, Cloudbreak UltraQ enables the development of highly sensitive assays.
ADD TO CART
Trinity-400.jpg
Trinity™ sequencing reagents simplify targeted sequencing by eliminating manual steps and automating key processes on the AVITI™ system. This streamlined approach delivers fast, high-quality and cost-effective results.
ADD TO CART
Resources
aviti-video-1-540.jpg
Introducing the all-in-one sequencing platform, with affordable quality for multi-scale applications.
Element-Biosciences-Video1-540.jpg
When you purchase an AVITI™ system, dedicated Field Application Scientists (FAS) are there to help guide you with customized training and ongoing support.
System-brochure-video-540.jpg
A detailed look at the Element AVITI platform and its low-throughput counterpart, the Element AVITI LT, engineered for reduced instrument cost.
System-specification-540.jpg
Learn how the Element AVITI™ system reimagines the core components of NGS to offer a benchtop platform that grants access to the genomics ecosystem.
Get the actionable insights for tomorrow’s breakthrough discoveries – today.
With independent flow cells that effectively offer the benefits of two sequencers in one, AVITI™ delivers exceptional data quality and industry-low sequencing costs, making high-performance sequencing both accessible and scalable.
TALK TO AN EXPERT