ContactUsLinkComponent
Ward's Plastinated Biological Specimens
Widest Selection of Permanently-Preserved Specimens
Ideally-suited for research, display, and hands-on demonstration, these actual biological specimens have been preserved and plastinated so they appear as vivid and life-like as their pre-preservation state, for years of student use.
  • Exclusive plastination process replaces water and fat with special reactive polymers
  • Long-lasting, fixed specimen position clearly shows organs and structures
  • Dry, odorless, and extremely durable
  • Largest selection of specimen options, only from Ward’s Science
plastinated-preserved-140.jpg
Shop for all Ward's plastinated biological specimens.