Orders LinkContactUsLinkComponent
Chemical Label Information | Avantor
Chemical Label Information
Every Ward's chemical label meets the latest OSHA/WHMIS labeling requirements in a clear and concise format and include the following information:
  • Product identifier - chemical name, grade and properties
  • Supplier identification - name, address and contact information
  • Pictograms - indicates specific information about chemical hazard(s)
  • Signal word - indicates severity of hazard(s)
  • Hazard statements - describes nature of hazard
  • Precautionary statements - recommended measures to prevent adverse effects
For help reading your Ward's chemical labels, or assistance and materials for updating your existing chemical labels to match the latest requirements, contact the Ward's Science Plus Us Chemistry team at [email protected].
ChemLabelInfo.jpg
See the information that must be on your chemical labels in order to be OSHA/WHMIS compliant.