Orders LinkContactUsLinkComponent
Avantor J.T.Baker® brand BAKERBOND® process chromatography resins comparison guide
For more than a century, customers around the world have trusted Avantor J.T.Baker® brand chemicals, reagents, excipients, chromatography resins and media to meet the needs of some of the most demanding applications.
Learn more about Avantor J.T.Baker® brand BAKERBOND® process chromatography resins and lab columns.
 Target applicationsFamilyFunctionalityNon-hazardous, non-flammable storage solutionAvailable in lab columnsAvailable as a bulk resin
J.T.Baker® BAKERBOND® 
PROchievA resin
The first, critical step in mAbs manufacturingAffinityHigh binding capacity resin with affinity to Fc region of mAbs
J.T.Baker® BAKERBOND® 
PolyABx
From mAbs to recombinant proteins and beyondIon Exchange (IEX)Multimode resin with primary weak cation exchanger with weak anion exchange sites+ hydrophilicity
J.T.Baker® BAKERBOND® 
PolyCSx
Multimode resin with primary strong cation exchanger with weak cation and anion exchange sites + hydrophilicity
J.T.Baker® BAKERBOND® 
PolyQUAT
Multimode resin with primary strong anion exchanger with weak anion exchange sites + hydrophilicity
J.T.Baker® BAKERBOND® 
PolyPEI
Multimode resin with primary weak anion exchanger + hydrophilicity
J.T.Baker® BAKERBOND® 
Poly Hi-Propyl
Hydrophobic Interaction (HIC)Mixed-mode resin with hydrophilicity + hydrophobicity