Orders LinkContactUsLinkComponent
hero blockprentpage.jpg
Comprehensive PFAS analysis solutions from Avantor®
Per- and polyfluoroalkyl substances (PFAS) have become a significant global environmental and public health issue because of their persistence, bioaccumulation and potential toxicity. More stringent regulatory requirements and increased research into PFAS have created an increased demand for accurate and reliable analytical solutions, including LC-MS grade PFAS free mobile phase additives of water, methanol, acetonitrile and ammonium acetate.
PFAS Analysis Solutions | Avantor Chromatography for CA Environmental Testing | Avantor
Empowering PFAS analysis
Avantor® works to empower scientists and researchers by providing them with the tools they need to meet the challenges of PFAS analysis. Our extensive portfolio of LC-MS grade PFAS free solvents and vials, caps, and kits for PFAS testing allow us to provide an end-to-end solution for detecting, quantifying and reporting PFAS across many applications.
Whether involved in environmental monitoring, food safety or human health impact studies, Avantor’s comprehensive range of high-performance instruments, consumables and expert support ensures you get the precision and reproducibility you need at every research stage. Avantor can be your trusted partner to advance PFAS research, meet regulatory demands and drive scientific progress in this critical field.
The importance of PFAS testing
PFAS compounds are neurotoxic, have been linked to disrupting reproduction and development and can harm humans and wildlife. PFAS can be found in many environments, including:
  • Drinking water
  • Soil and wastewater
  • Food and food packaging
  • Personal care products
  • Biosolids
That’s why accurately detecting and quantifying the presence of PFAS is critical to public and environmental safety.
Avantor’s solutions for PFAS analysis
Avantor offers a comprehensive range of PFAS analysis solutions, including LC-MS grade PFAS free solutions and additives like ammonium acetate, provide scientists with the necessary resources to generate reliable, high-quality data. Look at our PFAS portfolio, which includes everything you need, from advanced instruments to consumables, to support every phase of PFAS research.
Our solutions support key applications including:
  • Environmental testing: Soil, water, and air contamination
  • Food safety: Detecting PFAS in packaging and consumables
  • Human health: Assessing PFAS impacts on biological systems
undefined
Talk to a PFAS testing expert
Please contact us to speak with a member of our team.
Contact us
Featured Products
sampaling_treaser_image.jpg
A key requirement for PFAS testing is using PFAS-free container closure systems (CCS), which consist of a container and a sealing method. Avantor offers PFAS free vials and caps are available separately or as kits, providing flexibility for different testing needs. They are constructed using polypropylene (PP) which prevents PFAS contamination and are shipped in PFAS-free packaging.
mobile_preparation.jpg
Choosing the right chemicals is critical for chromatography, particularly in PFAS analysis. Avantor® offers a wide range of solvents and reagents that meet the strict requirements of PFAS analysis and work with many substances.
coloumnimage540x390.jpg
We offer a wide range of stationary phase selectivities in several formats for effective analyte separation in liquid and gas chromatography. Our GC and GC-MS columns are designed to meet your environmental application needs.
instrument_and_software.jpg
Avantor offers the instrumentation you need to reliably use high-performance liquid chromatography (HPLC) and , ultra-high-performance liquid chromatography and mass spectrometry in environmental analysis.
standaredsPFAS.jpg
High-quality reference materials from Avantor are critical for accurate analytical measurements and effective quality control, so you can be sure you have reliable data for sound decisions.
Chromatography Applications For PFAS Analysis
drinkingwaterEPA METHOD 537.1.jpg
Determination of selected PFAS in Drinking water by solid phase extraction and Liquid Chromatography/tandem mass spectrometry (LC-MS/MS).
Find the official method
Find our application notes
Drinking waterEPA METHOD 533.jpg
Determination of selected PFAS in Drinking water by Isotope dilution anion exchange solid phase extraction and Liquid Chromatography/tandem mass spectrometry (LC-MS/MS).
Find the official method
Find our application notes
wastwaterEPA METHOD 8327.jpg
Determination of selected PFAS in Surface water, Wastewater or groundwater by Liquid Chromatography/tandem mass spectrometry (LC-MS/MS).
Find the official method
Find our application notes
1fruitpictureEPA METHOD 1633.jpg
Determination of selected PFAS in aqueous, Solids, Biosolids and tissue samples by LC-MS/MS.
Find the official method
Find our application notes