Orders LinkContactUsLinkComponent
Solutionsa-for-research-1.jpg
Advanced Battery Science: Chemicals and Raw Materials for Production
Avantor® offers thousands of chemicals suitable for a multitude of analytical applications in R&D from our key brands – guaranteed high-quality chemicals and reproducible performance from batch to batch is key to achieving your QC, production or R&D goals.
Learn more
Chemicals and Raw Materials for production | Avantor
Featured Products and Services
anode1-90.jpg
Anodes are classically composed of natural or synthetic graphite, but researchers are investigating other carbon allotropes (carbon black, fullerenes) and non-carbon materials (lithium foil, oxides of bismuth, germanium, silicon and tin). Learn more.
cathode-100.jpg
Cathode materials typically include oxides of transition metals that can accommodate lithium by changing their oxidation states. We also offer many sodium-based and other salts for research into non-lithium technologies.
electrolyte-100.jpg
Conventional electrolytes consist of an organic solvent (such as ethylene carbonate) with a dissolved salt. Solid electrolytes have the potential to eliminate leakage and increase battery safety. See our product portfolio.
brinder-100.jpg
Separator materials used to control thermal conductance include polyethylene, polypropylene and other polymers in sheets or powder form.
metal-100.jpg
Pure metals and metal alloys used in batteries include aluminum, cobalt, stainless steel, and nickel in many forms, including foil, wires, powders and rods.
organic-100.jpg
A wide range of high purity solvents for battery production
cms-100.jpg
To help you better manage strategic priorities in your laboratory, we offer scaled volumes of custom formulated powders, solvents and solutions.
Questions about our offering of chemicals and raw materials for production?
Contact your local sales representative for more information or fill in our contact form.
Contact Us