PRIME-XV MSC Expansion XSFM is a serum-free and xeno-free complete medium optimized for the maintenance and expansion of purified human MSCs. This product does not contain antibiotics. PRIME-XV MSC Expansion XSFM is used in the maintenance and expans 1 * 1 L
Catalogus # SHBT91149-1L
Leverancier: Shenandoah Biotechnology
New Product
Frequently Bought Together