[EN]SPOUT FOR WIDE-NECK CANISTERS 1 * 5 ST
Catalogus # HUENET-281200
Leverancier: hünersdorff GmbH
New Product
[EN]SPOUT FOR WIDE-NECK CANISTERS 1 * 5 ST
Catalogus # HUENET-281200
Leverancier: hünersdorff GmbH
New Product
Frequently Bought Together
Pasteurpipetten
Leverancier: RATIOLAB
Aceton ≥99%, TECHNICAL
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverancier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Leverancier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Leverancier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Leverancier: VWR Chemicals
VWR®, Serologische pipetten, standaardreeks
Leverancier: VWR Collection
Pipetpunten, Finntip™
Leverancier: Thermo Fisher Scientific
VWR® Standaardlijntransferpipetten
Leverancier: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Leverancier: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
VWR®, Transferpipetten, Hoge Prestaties
Leverancier: VWR Collection
[EN]WIDE SCREW CONNECTION, HDPE, BLUE (F 1 * 2 ST
Leverancier: hünersdorff GmbH