Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Selenium(IV) oxide 99,8%

Selenium(IV) oxide 99,8%

Bottle
Catalogus # APOSIN9857-500G
Leverancier:   Apollo Scientific
Selenium(IV) oxide 99,8%
Catalogus # APOSIN9857-500G
Leverancier:  Apollo Scientific
CAS-nummer:  7446-08-4
Selenium(IV) oxide 99,8%

Bottle

CAS-nummer:  7446-08-4
Price
594,00 €
Log in voor beschikbaarheid
1 * 500 g
Product Details & Documents

Usage: Inorganic salt or catalyst

  • CAS-nummer:7446-08-4
  • Formule:H₂O₂Se
  • MDL-nummer:MFCD00003562
  • Molecular Weight:112,97467 g/mol
  • UN:3283
Specificaties

Specificaties

  • Verpakking:
    Bottle

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.