Fuchsin basic
Bottle
APOSBIGE1007-5G
Usage: Biochemical/ life science applications.
: CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
- :632-99-5
- :C₂₀H₁₉N₃·ClH
- :MFCD00012569
- :337,8459 g/mol
- :3077
- Verpakking:Bottle