[EN]JERRYCAN 10 L HDPE NATURAL W SCREWCA 1 * 10 ST
Catalog # RIXI6020010045WC
Supplier: RIXIUS
New Product
[EN]JERRYCAN 10 L HDPE NATURAL W SCREWCA 1 * 10 ST
Catalog # RIXI6020010045WC
Supplier: RIXIUS
New Product
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Shoe covers, PP/CPE
Supplier: VWR Collection
VWR®, Jerrycans, brede hals, met schroefdoppen
Supplier: VWR Collection
VWR®, Nitrilhandschoenen
Supplier: VWR Collection
Flessen, brede hals, vierkant zonder schroefdop
Supplier: Kautex
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Gelladende Pipettips
Supplier: VWR Collection
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
VWR®, Zakken voor eenmalig gebruik
Supplier: VWR Collection
VWR®, Transferpipetten, Hoge Prestaties
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Citroenzuur
Supplier: Hach
Jerricans, Narrow Neck, HDPE
Supplier: RIXIUS