Orders LinkContactUsLinkComponent
Antibiotic Sensitivity Discs
Antibiotic Sensitivity Discs
Doxycycline hydrochloride, 30 mcg, 50
Catalogus # HIMESD012-5X50DS
Leverancier:   HIMEDIA
Antibiotic Sensitivity Discs
Catalogus # HIMESD012-5X50DS
Leverancier:  HIMEDIA
CAS-nummer:  
Antibiotic Sensitivity Discs
Doxycycline hydrochloride, 30 mcg, 50
CAS-nummer:  
Price
19,50 €
Log in voor beschikbaarheid
1 * 50 ST
Product Details & Documents
Antibiotic sensitivity disc are used to determine the effectivness of an antibiotic against a microorganism as a guide to antibiotic therapy.
The disc consist of a specific concentration of an antibiotic in mcg which is tested against a microorganism of a standard density, swabbed uniformly on an appropriate media following overnight incubation.
The antibiotic is impregnated on a filter paper disc in specified concentration. Both sides of the disc are printed with name and concentration for easy identification.
Specificaties
Specificaties
  • VPE:
    50
  • Omschrijving:
    Doxycycline hydrochloride, 30 mcg