Orders LinkContactUsLinkComponent
Toebehoren
Milli-Q® SP 240 prefilter module, for pretreatment of potable tap water to feed the pure RO water production station, includes a pressure regulator, requires one power cord
Catalogus # MILLZSR240PFT0
Leverancier:   Merck
Toebehoren
Catalogus # MILLZSR240PFT0
Leverancier:  Merck
Toebehoren
Milli-Q® SP 240 prefilter module, for pretreatment of potable tap water to feed the pure RO water production station, includes a pressure regulator, requires one power cord
Price
1 580,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Specificaties
  • Omschrijving:
    Milli-Q® SP 240 prefilter module, for pretreatment of potable tap water to feed the pure RO water production station, includes a pressure regulator, requires one power cord
  • Seq:
    50