Flasks, round bottom, with three parallel necks
100 ml, 14/23 NS, 14/23 NS
Catalogus # NEUB0760-0100-14
Leverancier: NEUBERT VOLUME GLASSWAERE
Flasks, round bottom, with three parallel necks
Catalogus # NEUB0760-0100-14
Leverancier: NEUBERT VOLUME GLASSWAERE
Ga naar:
Product Details & Documents
Borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
Specificaties
- Capaciteit:100 ml
- Seq:02
- Zijhalzen:14/23 NS
- Middenhals:14/23 NS