Safety spectacles, Slam
Anti-scratch, zwart, Green PC
Catalogus # BOLESLAWPCC5
Leverancier: Bollé Safety
Ga naar:
Product Details & Documents
Modern design spectacles with extra-easy fitting and comfort.
- Panoramic visual field
- Anti-scratch, anti-fog, anti-static coatings
- Ultra enveloping
- Comfort temples
Weight: 23 g.
Certifications: EN 166, EN 170 (clear lens), EN 172 (smoke lens).
Delivery information: Supplied with adjustable cord.
Specificaties
- Type:Anti-scratch
- Kleur:zwart
- Seq:03
- Beglazing:Green PC