Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Safety spectacles, Slam
Safety spectacles, Slam
Anti-scratch, zwart, Green PC
Catalogus # BOLESLAWPCC5
Leverancier:   Bollé Safety
Safety spectacles, Slam
Catalogus # BOLESLAWPCC5
Leverancier:  Bollé Safety
Safety spectacles, Slam
Safety spectacles, Slam
Anti-scratch, zwart, Green PC
Price
13,10 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Modern design spectacles with extra-easy fitting and comfort.
  • Panoramic visual field
  • Anti-scratch, anti-fog, anti-static coatings
  • Ultra enveloping
  • Comfort temples
Weight: 23 g.
Certifications: EN 166, EN 170 (clear lens), EN 172 (smoke lens).
Delivery information: Supplied with adjustable cord.
Specificaties
  • Type:
    Anti-scratch
  • Kleur:
    zwart
  • Seq:
    03
  • Beglazing:
    Green PC