Nunc™ Universal Tri-coded Tubes
Externally-threaded universal tube, cylindrical, graduated, 1,80 ml, Kunststof, +, Rack, Case of 480, 10 bags of 48 tubes
Catalogus # 479-2517
Leverancier: Thermo Fisher Scientific
Nunc™ Universal Tri-coded Tubes
Catalogus # 479-2517
Leverancier: Thermo Fisher Scientific
Ga naar:
Product Details & Documents
Nunc™ Universal Tri-coded Tubes to improve the efficiency and throughput of your sample storage workflow.
- Tri-coded tubes come sterile and offered in both racked and bulk formats
- The 2D screwtop tubes meet a range of manual and automated long term storage needs
- It can be used in combination with Thermo Scientific barcode readers, arrayers and capping equipment
Tubes are molded from medical grade resins and feature a permanent, high contrast 2D, 1D linear and human readable barcode.
Specificaties
- Verpakt:Rack, Case of 480, 10 bags of 48 tubes
- Steriel:+
- Capaciteit:1,80 ml
- Omschrijving:Externally-threaded universal tube, cylindrical, graduated
- Materiaal:Kunststof
- Seq:02
- Cat. No.:479-2517
- Milieu certificering:NAR Certificate of Retirement
- Environmentally Preferable:Lage impact op de fabricage



