Natriumhydroxide 15% in aqueous solution
−, 500 mL
Leverancier: PROLABO CMP
Natriumhydroxide 15% in aqueous solution
Catalogus # PRLSMC311096.0500
Leverancier: PROLABO CMP
CAS-nummer: 1310-73-2
Ga naar:
Product Details & Documents
Synoniemen: Caustic soda
- CAS-nummer:1310-73-2
- Formule:NaOH
- MDL-nummer:MFCD00003548
- Molecular Weight:40 g/mol
- Kokend Pt:119…135 °C (1013 hPa)
- Smelten Pt:9…10 °C
- Dichtheid:1,53 g/cm³ (20 °C)
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:215-185-5
- UN:1824
- ADR:8,II
Specificaties
Specificaties
- Verpakking:−
- VPE:500 mL
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



