Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Glass inserts, AQ™

Glass inserts, AQ™

Glass inserts, low volume, with plastic spring attached. For use in 2 ml autosampler vials, 0,20 ml, Conical bottom, 6×29 mm, Glas, Neen
Catalogus # MICS9502S-02E
Leverancier:   MICROSOLV
Glass inserts, AQ™
Catalogus # MICS9502S-02E
Leverancier:  MICROSOLV
Glass inserts, AQ™
Glass inserts, AQ™

Glass inserts, low volume, with plastic spring attached. For use in 2 ml autosampler vials, 0,20 ml, Conical bottom, 6×29 mm, Glas, Neen

Price
149,00 €
Log in voor beschikbaarheid
1 * 100 ST
Product Details & Documents

These vial inserts are made from borosilicate glass.

Specificaties
  • Inclusief dop:
    Neen
  • Ø×h:
    6×29 mm
  • Capaciteit:
    0,20 ml
  • Omschrijving:
    Glass inserts, low volume, with plastic spring attached. For use in 2 ml autosampler vials
  • Materiaal:
    Glas
  • Seq:
    06
  • Bodem:
    Conical bottom