Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Plunger cans

Plunger cans

Plunger Can, 1 l, 120×115×265 mm
Catalogus # MSGA40097
Leverancier:   asecos
Plunger cans
Catalogus # MSGA40097
Leverancier:  asecos
Plunger cans
Plunger cans

Plunger Can, 1 l, 120×115×265 mm

Price
99,80 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

These plunger cans are dedicated to the safe moistening of cloth, wipes and tissues.

  • Unused liquid flows back into the container
  • Spring mounted plate includes flame barrier
  • Shock resistant and explosion proof (PE models only)
  • Cans with protector ring (stainless steel models only)
Specificaties
  • Capaciteit:
    1 l
  • Omschrijving:
    Plunger Can
  • Seq:
    11
  • b×d×h:
    120×115×265 mm